siRNA Details
Gene Symbol: Atp6v0d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011927.1 | NP_001011927.1 | 2, 3 | 390 | ||
GenBank mRNA | BC088462.1 | - | 2, 3 | 390 | 7127604 | |
FQ213716.1 | - | 2, 3 | 406 | |||
FQ215412.1 | - | 1, 2 | 125 |
