siRNA Details
Gene Symbol: Atp6v0d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011927.1 | NP_001011927.1 | 2 | 282 | ||
GenBank mRNA | BC088462.1 | - | 2 | 282 | 7127604 | |
FQ213716.1 | - | 2 | 298 | |||
FQ215412.1 | - | 1 | 17 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_013477, AK083372, U13840, AB088408, AK171515, and others.
Gene Ontology Categories:
