siRNA Details
Gene Symbol: Nutf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007629.1 | NP_001007630.1 | 2 | 118 | ||
XM_006255480.2 | XP_006255542.1 | 1 | 82 | |||
GenBank mRNA | BC061569.1 | - | 2 | 118 | 6886638 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
