siRNA Details
Gene Symbol: Psmb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025637.1 | NP_001020808.1 | 7 | 815 | ||
GenBank mRNA | BC098835.1 | - | 7 | 815 | 7378841 | |
FQ217478.1 | - | 3 | 358 | |||
FQ226463.1 | - | 7 | 808 | |||
FQ228226.1 | - | 7 | 795 | |||
FQ229295.1 | - | 7 | 813 | |||
FQ233484.1 | - | 7 | 825 | |||
FQ234080.1 | - | 7 | 817 | |||
FQ235075.1 | - | 7 | 798 |
