siRNA Details
Gene Symbol: Dpep2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011928.1 | NP_001011928.1 | 11 | 1466 | ||
XM_008772487.1 | XP_008770709.1 | 11 | 1763 | |||
GenBank mRNA | BC088195.1 | - | 11 | 1466 | 7374282 |
