siRNA Details
Gene Symbol: Dhx38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106185.1 | NP_001099655.1 | 12 | 1892 | ||
XM_006255577.2 | XP_006255639.1 | 12 | 1938 | |||
XM_008772489.1 | XP_008770711.1 | 12 | 2481 |
