siRNA Details
Gene Symbol: Txnl4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013891.1 | NP_001013913.1 | 3 | 445 | ||
XM_006255579.2 | XP_006255641.1 | 4 | 555 | |||
GenBank mRNA | BC089962.1 | - | 3 | 445 | 7314335 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
