siRNA Details
Gene Symbol: Sf3b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106187.2 | NP_001099657.2 | 22 | 3201 | ||
XM_006255620.2 | XP_006255682.1 | 22 | 3336 | |||
GenBank mRNA | BC168883.1 | - | 16 | 2106 |

More Information
Other Species Targeted

Gene Ontology Categories:
