siRNA Details
Gene Symbol: Banp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106191.1 | NP_001099661.1 | 5 | 536 | ||
XM_006255751.2 | XP_006255813.1 | 5 | 582 | |||
XM_006255752.2 | XP_006255814.1 | 5 | 581 | |||
XM_006255755.2 | XP_006255817.1 | 5 | 581 | |||
XM_006255756.2 | XP_006255818.1 | 5 | 581 | |||
XM_008772601.1 | XP_008770823.1 | 5 | 803 | |||
XM_008772602.1 | XP_008770824.1 | 5 | 582 | |||
GenBank mRNA | BC160844.1 | - | 5 | 563 | 8361869 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_016812, NM_001110100, AK184999, BC062641, BC021650, and others.
Gene Ontology Categories:
