siRNA Details
Gene Symbol: Banp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106191.1 | NP_001099661.1 | 12, 13 | 1577 | ||
XM_006255751.2 | XP_006255813.1 | 12, 13 | 1623 | |||
XM_006255752.2 | XP_006255814.1 | 12, 13 | 1613 | |||
XM_006255753.2 | XP_006255815.1 | 11, 12 | 1505 | |||
XM_006255754.2 | XP_006255816.1 | 11, 12 | 1496 | |||
XM_008772601.1 | XP_008770823.1 | 12, 13 | 1844 | |||
GenBank mRNA | BC160844.1 | - | 12, 13 | 1604 | 8361869 |
