siRNA Details
Gene Symbol: Nup133 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772670.1 | XP_008770892.1 | 25 | 3425 | ||
XM_008772671.1 | XP_008770893.1 | 25 | 3544 |

More Information
Other Species Targeted

Gene Ontology Categories:
