siRNA Details
Gene Symbol: Ttc13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136162.1 | NP_001129634.1 | 5 | 517 | ||
XM_006255802.2 | XP_006255864.1 | 7 | 685 | |||
XM_006255803.2 | XP_006255865.1 | 7 | 685 | |||
XM_006255805.2 | XP_006255867.1 | 5 | 526 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
