siRNA Details
Gene Symbol: Tdrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106202.1 | NP_001099672.1 | 23 | 3587 | ||
XM_006231642.2 | XP_006231704.1 | 24 | 3850 | |||
XM_008760524.1 | XP_008758746.1 | 24 | 3657 | |||
XM_008760525.1 | XP_008758747.1 | 24 | 3622 | |||
XM_008760526.1 | XP_008758748.1 | 23 | 3787 | |||
XM_008760528.1 | XP_008758750.1 | 23 | 3416 | |||
XM_008760529.1 | XP_008758751.1 | 23 | 3694 | |||
XM_008760530.1 | XP_008758752.1 | 23 | 3607 | |||
XM_008760531.1 | XP_008758753.1 | 22 | 3436 |
