siRNA Details
Gene Symbol: Sh3glb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011929.1 | NP_001011929.1 | 7 | 1039 | ||
XM_006233431.2 | XP_006233493.1 | 9 | 1165 | |||
XM_006233433.2 | XP_006233495.1 | 9 | 1115 | |||
XM_006233435.2 | XP_006233497.1 | 7 | 1051 | |||
GenBank mRNA | BC079085.1 | - | 7 | 1039 | 7123144 | |
FQ216102.1 | - | 3 | 253 | |||
FQ234013.1 | - | 7 | 762 |
