siRNA Details
Gene Symbol: Eya4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271317.1 | NP_001258246.1 | 15, 16 | 1590 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_010167, AK041625, BC120899, BC120898, Y17115.
Gene Ontology Categories:
