siRNA Details
Gene Symbol: Eya4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271317.1 | NP_001258246.1 | 12 | 1253 | ||
XM_008758600.1 | XP_008756822.1 | 12 | 1115 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Eya4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271317.1 | NP_001258246.1 | 12 | 1253 | ||
XM_008758600.1 | XP_008756822.1 | 12 | 1115 |