siRNA Details
Gene Symbol: Eya4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271317.1 | NP_001258246.1 | 17 | 1843 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_010167, AJ007997, AK041625, BC120899, BC120898, and others.
Gene Ontology Categories:
