siRNA Details
Gene Symbol: RGD1305235 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017447.1 | NP_001017447.1 | 5 | 759 | ||
XM_006227833.2 | XP_006227895.1 | 4 | 897 | |||
GenBank mRNA | BC078920.1 | - | 5 | 759 | 7096641 |
