siRNA Details
Gene Symbol: RGD1305235 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017447.1 | NP_001017447.1 | 2 | 147 | ||
GenBank mRNA | BC078920.1 | - | 2 | 147 | 7096641 |

More Information
Other Species Targeted

