siRNA Details
Gene Symbol: Aig1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134425.1 | NP_001127897.1 | 5 | 576 | ||
XM_006227642.2 | XP_006227704.1 | 5 | 775 | |||
GenBank mRNA | BC167098.1 | - | 5 | 576 | 7319120 | |
FQ228880.1 | - | 5 | 648 | |||
FQ229550.1 | - | 5 | 594 | |||
FQ229983.1 | - | 5 | 592 |

More Information
Other Species Targeted

