siRNA Details
Gene Symbol: Aurkc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758900.1 | XP_008757122.1 | 3 | 241 | ||
XM_008758901.1 | XP_008757123.1 | 4 | 344 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
