siRNA Details
Gene Symbol: Ttyh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106225.1 | NP_001099695.1 | 7, 8 | 983 | ||
XM_006228216.2 | XP_006228278.1 | 7, 8 | 1007 | |||
GenBank mRNA | BC168158.1 | - | 7, 8 | 978 | 8364284 | |
FQ212233.1 | - | 4, 5 | 448 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
