siRNA Details
Gene Symbol: Gltscr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106226.2 | NP_001099696.2 | 10 | 3377 | ||
XM_006228343.2 | XP_006228405.1 | 12 | 3724 | |||
XM_006228344.2 | XP_006228406.1 | 11 | 3658 | |||
XM_006228345.2 | XP_006228407.1 | 11 | 3547 |

More Information
Other Species Targeted

