siRNA Details
Gene Symbol: Gltscr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106226.2 | NP_001099696.2 | 2, 3 | 85 | ||
XM_006228343.2 | XP_006228405.1 | 4, 5 | 432 | |||
XM_006228345.2 | XP_006228407.1 | 4, 5 | 432 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
