siRNA Details
Gene Symbol: Gltscr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106226.2 | NP_001099696.2 | 12 | 3589 | ||
XM_006228343.2 | XP_006228405.1 | 14 | 3936 | |||
XM_006228344.2 | XP_006228406.1 | 13 | 3870 | |||
XM_006228345.2 | XP_006228407.1 | 13 | 3759 |

More Information
Other Species Targeted

