siRNA Details
Gene Symbol: Prkd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013895.1 | NP_001013917.1 | 16 | 2407 | ||
XM_008758905.1 | XP_008757127.1 | 16 | 3294 | |||
GenBank mRNA | BC083592.1 | - | 16 | 2407 | 7132731 | |
FQ218070.1 | - | 17 | 2498 |
