siRNA Details
Gene Symbol: Psg29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025641.1 | NP_001020812.1 | 5 | 1361 | ||
GenBank mRNA | BC097354.1 | - | 5 | 1361 | 7441714 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
