siRNA Details
Gene Symbol: Psg29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025641.1 | NP_001020812.1 | 2 | 476 | ||
GenBank mRNA | BC097354.1 | - | 2 | 476 | 7441714 |

More Information
Other Species Targeted

Gene Ontology Categories:
