siRNA Details
Gene Symbol: APLP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142276.1 | NP_001135748.1 | 6 | 972 | ||
NM_001142277.1 | NP_001135749.1 | 6 | 972 | |||
NM_001243299.1 | NP_001230228.1 | 6 | 890 | |||
NM_001642.2 | NP_001633.1 | 6 | 972 | |||
NR_024515.1 | 4 | 638 | ||||
NR_024516.1 | 4 | 674 | ||||
GenBank mRNA | AF168956.1 | - | 6 | 818 | ||
AK056883.1 | - | 4 | 565 | |||
AK128162.1 | - | 6 | 883 | |||
AK225283.1 | - | 6 | 899 | |||
AK300378.1 | - | 4 | 601 | |||
AK304736.1 | - | 6 | 881 | |||
AK308248.1 | - | 4 | 516 | |||
BC000373.2 | - | 6 | 872 | 2820109 | ||
L09209.1 | - | 6 | 887 | |||
L19597.1 | - | 6 | 883 | |||
L23113.1 | - | 2 | 133 | |||
L23114.1 | - | 5 | 703 | |||
L27631.1 | - | 6 | 888 | |||
S60099.1 | - | 6 | 887 | |||
Z22572.1 | - | 6 | 882 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
