siRNA Details
Gene Symbol: Fbxo46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025642.1 | NP_001020813.1 | 2 | 1864 | ||
XM_006228399.2 | XP_006228461.1 | 3 | 2051 | |||
XM_006228400.2 | XP_006228462.1 | 3 | 2663 | |||
XM_006228401.2 | XP_006228463.1 | 2 | 2546 | |||
XM_008758910.1 | XP_008757132.1 | 2 | 2195 | |||
GenBank mRNA | BC098945.1 | - | 2 | 1864 | 7458607 |

More Information
Other Species Targeted

