siRNA Details
Gene Symbol: Ethe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106234.1 | NP_001099704.1 | 6 | 701 | ||
GenBank mRNA | BC158848.1 | - | 6 | 661 | 7444454 | |
FQ209618.1 | - | 6 | 672 | |||
FQ221207.1 | - | 2 | 113 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
