siRNA Details
Gene Symbol: Snrpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008303.1 | NP_001008304.1 | 3 | 313 | ||
XM_006228564.2 | XP_006228626.1 | 3 | 444 | |||
GenBank mRNA | BC086331.1 | - | 3 | 313 | 7300752 | |
FQ227089.1 | - | 2 | 207 | |||
FQ231860.1 | - | 1 | 17 | |||
FQ233559.1 | - | 2 | 207 |

More Information
Other Species Targeted

Gene Ontology Categories:
