siRNA Details
Gene Symbol: Ltbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170336.1 | NP_001163807.1 | 13, 14 | 1778 | ||
XM_006228570.2 | XP_006228632.1 | 10, 11 | 1620 | |||
XM_008759116.1 | XP_008757338.1 | 13, 14 | 1685 |
