siRNA Details
Gene Symbol: Plekhg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134972.1 | NP_001128444.1 | 19 | 4292 | ||
XM_006228613.2 | XP_006228675.1 | 17 | 4138 | |||
XM_006228614.1 | XP_006228676.1 | 18 | 4045 | |||
XM_006228615.2 | XP_006228677.1 | 19 | 4981 | |||
XM_006228616.2 | XP_006228678.1 | 19 | 3734 | |||
XM_006228617.2 | XP_006228679.1 | 18 | 3781 | |||
XM_006228618.2 | XP_006228680.1 | 18 | 3778 | |||
XM_006228619.2 | XP_006228681.1 | 18 | 3736 | |||
XM_006228620.2 | XP_006228682.1 | 18 | 3733 |
