siRNA Details
Gene Symbol: Fbxo17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013064.2 | NP_001013082.2 | 6 | 909 | ||
GenBank mRNA | BC079218.1 | - | 6 | 909 | 7131902 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
