siRNA Details
Gene Symbol: Rinl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106241.1 | NP_001099711.1 | 8 | 820 | ||
XM_006228653.2 | XP_006228715.1 | 8 | 1072 | |||
XM_006228654.2 | XP_006228716.1 | 8 | 1060 | |||
XM_006228655.2 | XP_006228717.1 | 8 | 1072 | |||
XM_006228656.2 | XP_006228718.1 | 8 | 1072 | |||
XM_006228657.2 | XP_006228719.1 | 7 | 795 | |||
XM_008759120.1 | XP_008757342.1 | 7 | 937 | |||
XM_008759121.1 | XP_008757343.1 | 8 | 905 |
