siRNA Details
Gene Symbol: Eif3k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006228715.2 | XP_006228777.1 | 3, 4 | 373 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001285943, NM_001285942, NM_028659, AK037834, AK158875, and others.
Gene Ontology Categories:
