siRNA Details
Gene Symbol: Map4k1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106243.1 | NP_001099713.1 | 1 | 146 | ||
XM_006228716.2 | XP_006228778.1 | 1 | 1108 | |||
XM_008759122.1 | XP_008757344.1 | 1 | 1108 |

More Information
Other Species Targeted

Gene Ontology Categories:
