siRNA Details
Gene Symbol: Psmd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100831.1 | NP_001094301.1 | 3 | 566 | ||
GenBank mRNA | BC105894.1 | - | 3 | 331 | ||
CD371338.1 | - | 3 | 576 | |||
FQ213223.1 | - | 3 | 366 | |||
FQ214314.1 | - | 3 | 363 | |||
FQ223219.1 | - | 3 | 409 | |||
FQ233693.1 | - | 3 | 397 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_026545, AK150493, AK168886, AK159795, AK027973, and others.
Gene Ontology Categories:
