siRNA Details
Gene Symbol: Psmd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100831.1 | NP_001094301.1 | 1 | 358 | ||
GenBank mRNA | BC105894.1 | - | 1 | 123 | ||
CD371338.1 | - | 1 | 368 | |||
FQ213223.1 | - | 1 | 158 | |||
FQ214314.1 | - | 1 | 155 | |||
FQ223219.1 | - | 1 | 201 | |||
FQ233693.1 | - | 1 | 189 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
