siRNA Details
Gene Symbol: Tbcb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040180.1 | NP_001035270.1 | 4 | 619 | ||
GenBank mRNA | AB256046.1 | - | 4 | 619 | ||
AM258961.1 | - | 4 | 619 | |||
BC126061.1 | - | 4 | 437 | 7113234 | ||
DQ447885.1 | - | 4 | 418 |
Gene Symbol: LOC103690005 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759264.1 | XP_008757486.1 | 4 | 449 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
