siRNA Details
Gene Symbol: RGD1560986 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759244.1 | XP_008757466.1 | 9 | 1770 | ||
XM_008759245.1 | XP_008757467.1 | 9 | 1770 | |||
XM_008759246.1 | XP_008757468.1 | 10 | 1695 | |||
XM_008759247.1 | XP_008757469.1 | 10 | 1695 | |||
XM_008759248.1 | XP_008757470.1 | 11 | 1620 | |||
XM_008759249.1 | XP_008757471.1 | 9 | 1770 | |||
XM_008759250.1 | XP_008757472.1 | 9 | 1770 | |||
XM_008759251.1 | XP_008757473.1 | 9 | 1770 | |||
XM_008759252.1 | XP_008757474.1 | 9 | 1770 |
