siRNA Details
Gene Symbol: Lin37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106245.1 | NP_001099715.1 | 9 | 865 | ||
XM_006228769.2 | XP_006228831.1 | 9 | 1286 | |||
GenBank mRNA | FQ233946.1 | - | 9 | 833 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
