siRNA Details
Gene Symbol: Tmem147 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038494.1 | NP_001033583.1 | 6 | 576 | ||
GenBank mRNA | BC111082.1 | - | 6 | 576 | 7455198 | |
FQ234356.1 | - | 6 | 591 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_027215, AK010168, BC147301, AK008089, BC147300, and others.
