siRNA Details
Gene Symbol: Ffar2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005877.1 | NP_001005877.1 | 1 | 228 | ||
XM_006228821.2 | XP_006228883.1 | 2 | 980 | |||
XM_006228822.2 | XP_006228884.1 | 3 | 574 | |||
XM_008759133.1 | XP_008757355.1 | 3 | 674 | |||
XM_008759134.1 | XP_008757356.1 | 3 | 635 | |||
XM_008759135.1 | XP_008757357.1 | 3 | 352 | |||
GenBank mRNA | AB106675.1 | - | 1 | 228 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
