siRNA Details
Gene Symbol: LOC100912106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748838.3 | XP_003748886.1 | 5 | 411 | ||
XM_006228897.2 | XP_006228959.1 | 5 | 275 |
Gene Symbol: Pdcd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106247.1 | NP_001099717.1 | 5 | 407 | ||
XM_006228884.2 | XP_006228946.1 | 5 | 275 | |||
GenBank mRNA | FQ222074.1 | - | 5 | 322 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
