siRNA Details
Gene Symbol: Zfp507 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106248.1 | NP_001099718.2 | 3 | 1517 | ||
XM_006228885.2 | XP_006228947.1 | 3 | 1478 | |||
XM_006228886.2 | XP_006228948.1 | 3 | 1549 |
Gene Symbol: LOC100912350 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748841.3 | XP_003748889.1 | 1980 | |||
XM_006228898.2 | XP_006228960.1 | 1980 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
