siRNA Details
Gene Symbol: Zfp536 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759267.1 | XP_008757489.1 | 6 | 4058 | ||
XM_008759268.1 | XP_008757490.1 | 6 | 3893 | |||
XM_008759269.1 | XP_008757491.1 | 6 | 3852 | |||
XM_008759270.1 | XP_008757492.1 | 6 | 3654 | |||
XM_008759271.1 | XP_008757493.1 | 6 | 3817 | |||
XM_008759272.1 | XP_008757494.1 | 6 | 3108 | |||
XM_008759273.1 | XP_008757495.1 | 6 | 3112 | |||
XM_008759274.1 | XP_008757496.1 | 6 | 3693 | |||
XM_008759275.1 | XP_008757497.1 | 6 | 3210 |
