siRNA Details
Gene Symbol: Ctu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106251.1 | NP_001099721.1 | 3 | 1827 | ||
XM_006228984.2 | XP_006229046.1 | 4 | 2071 | |||
XM_008759330.1 | XP_008757552.1 | 3 | 3164 | |||
GenBank mRNA | BC161898.1 | - | 3 | 1810 | 8367021 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
