siRNA Details
Gene Symbol: Klk10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004100.1 | NP_001004100.1 | 3 | 426 | ||
XM_006228987.2 | XP_006229049.1 | 4 | 561 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Klk10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004100.1 | NP_001004100.1 | 3 | 426 | ||
XM_006228987.2 | XP_006229049.1 | 4 | 561 |